Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCCATTAAATAAATGATCCAG[G/T]GAGTATATGGATCTTAACAAATGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600839 | ||||||||||||||||||||
Literature Links: |
CTXN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTXN2 - cortexin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145668.1 | Intron | NP_001139140.1 | ||||
XM_005254378.3 | Intron | XP_005254435.1 | ||||
XM_006720504.2 | Intron | XP_006720567.1 | ||||
XM_011521553.1 | Intron | XP_011519855.1 | ||||
XM_017022178.1 | Intron | XP_016877667.1 |
SLC12A1 - solute carrier family 12 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |