Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613639 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRD1 - adhesion G protein-coupled receptor D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005253566.2 | Intron | XP_005253623.1 | ||||
XM_011538203.2 | Intron | XP_011536505.1 | ||||
XM_011538204.2 | Intron | XP_011536506.1 | ||||
XM_011538205.2 | Intron | XP_011536507.1 | ||||
XM_011538206.2 | Intron | XP_011536508.1 | ||||
XM_011538207.2 | Intron | XP_011536509.1 | ||||
XM_011538208.2 | Intron | XP_011536510.1 | ||||
XM_011538209.2 | Intron | XP_011536511.1 | ||||
XM_011538210.2 | Intron | XP_011536512.1 | ||||
XM_011538211.2 | Intron | XP_011536513.1 | ||||
XM_011538212.2 | Intron | XP_011536514.1 |