Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTAAACAAACAAGTAGCAAATTT[A/G]GGTTTTGTTAATTTAGCTTTTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610640 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
YTHDF2 - YTH N6-methyladenosine RNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172828.1 | Intron | NP_001166299.1 | ||||
NM_001173128.1 | Intron | NP_001166599.1 | ||||
NM_016258.2 | Intron | NP_057342.2 |