Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTCGTCACTTCTTCTGTCAATC[A/G]GGTCCTCATTTGCCCTTTGCGTGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613570 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDAH PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LDAH - lipid droplet associated hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282719.1 | Intron | NP_001269648.1 | ||||
NM_001282720.1 | Intron | NP_001269649.1 | ||||
NM_001282721.1 | Intron | NP_001269650.1 | ||||
NM_001282722.1 | Intron | NP_001269651.1 | ||||
NM_001282723.1 | Intron | NP_001269652.1 | ||||
NM_001282724.1 | Intron | NP_001269653.1 | ||||
NM_021925.3 | Intron | NP_068744.1 | ||||
XM_011533014.2 | Intron | XP_011531316.1 |
Set Membership: |
HapMap |