Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCATACCTGAAAATTTTCAACAA[C/T]TAGCAGTATCATCAGAATCTTATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605103 MIM: 611676 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMU PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NMU - neuromedin U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292045.1 | Intron | NP_001278974.1 | ||||
NM_001292046.1 | Intron | NP_001278975.1 | ||||
NM_006681.3 | Intron | NP_006672.1 | ||||
XM_011534367.2 | Intron | XP_011532669.1 | ||||
XM_011534368.2 | Intron | XP_011532670.1 |
PDCL2 - phosducin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |