Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGAGGTATTTAAAGTGTTCTTT[A/G]AGCAGTTTTAATGCTATTCTTTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
71 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602105 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ASB17 - ankyrin repeat and SOCS box containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080868.2 | Intron | NP_543144.1 | ||||
XM_006710345.3 | Intron | XP_006710408.1 | ||||
XM_006710346.3 | Intron | XP_006710409.1 |
MSH4 - mutS homolog 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |