Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185430 MIM: 132811 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLU PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLU - clusterin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001831.3 | 1852 | UTR 3 | NP_001822.3 | |||
XM_006716284.2 | 1852 | Intron | XP_006716347.1 |
EPHX2 - epoxide hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256482.1 | 1852 | Intron | NP_001243411.1 | |||
NM_001256483.1 | 1852 | Intron | NP_001243412.1 | |||
NM_001256484.1 | 1852 | Intron | NP_001243413.1 | |||
NM_001979.5 | 1852 | Intron | NP_001970.2 | |||
XM_017013199.1 | 1852 | Intron | XP_016868688.1 | |||
XM_017013200.1 | 1852 | Intron | XP_016868689.1 |
Set Membership: |
HapMap |