Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTTTGATATGAAACATATACA[A/G]GCACACCTCAGAGATATTTCAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHS1 - pleckstrin homology domain containing S1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193434.1 | Intron | NP_001180363.1 | ||||
NM_001193435.1 | Intron | NP_001180364.1 | ||||
NM_024889.4 | Intron | NP_079165.3 | ||||
NM_182601.1 | Intron | NP_872407.1 | ||||
XM_005270162.2 | Intron | XP_005270219.1 | ||||
XM_005270163.2 | Intron | XP_005270220.1 | ||||
XM_011540171.1 | Intron | XP_011538473.1 | ||||
XM_011540172.1 | Intron | XP_011538474.1 | ||||
XM_011540173.1 | Intron | XP_011538475.1 | ||||
XM_011540174.2 | Intron | XP_011538476.1 | ||||
XM_011540175.1 | Intron | XP_011538477.1 | ||||
XM_011540176.1 | Intron | XP_011538478.1 | ||||
XM_017016666.1 | Intron | XP_016872155.1 |