Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL26A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL26A1 - collagen type XXVI alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278563.2 | 584 | UTR 5 | NP_001265492.1 | |||
NM_133457.4 | 584 | UTR 5 | NP_597714.2 | |||
XM_017011743.1 | 584 | UTR 5 | XP_016867232.1 | |||
XM_017011744.1 | 584 | Intron | XP_016867233.1 | |||
XM_017011745.1 | 584 | Intron | XP_016867234.1 |