Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607506 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAMTS14 - ADAM metallopeptidase with thrombospondin type 1 motif 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080722.4 | Intron | NP_542453.2 | ||||
NM_139155.3 | Intron | NP_631894.2 | ||||
XM_011539300.2 | Intron | XP_011537602.1 | ||||
XM_011539301.2 | Intron | XP_011537603.1 | ||||
XM_011539302.2 | Intron | XP_011537604.1 | ||||
XM_011539303.2 | Intron | XP_011537605.1 | ||||
XM_011539305.2 | Intron | XP_011537607.1 | ||||
XM_011539306.2 | Intron | XP_011537608.1 | ||||
XM_011539307.2 | Intron | XP_011537609.1 | ||||
XM_011539308.2 | Intron | XP_011537610.1 | ||||
XM_011539309.1 | Intron | XP_011537611.1 |
Set Membership: |
HapMap |