Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCACACAAACAGAAAGCATGTC[C/T]TTTAATTTTACCTATCCTTCAAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130590 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EEF1A1 - eukaryotic translation elongation factor 1 alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001402.5 | 2632 | UTR 3 | NP_001393.1 | |||
XM_011535514.2 | 2632 | Intron | XP_011533816.1 |
Set Membership: |
HapMap |