Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612990 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASXL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ASXL1 - additional sex combs like 1, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164603.1 | 1226 | Intron | NP_001158075.1 | |||
NM_015338.5 | 1226 | Intron | NP_056153.2 | |||
XM_006723727.3 | 1226 | Intron | XP_006723790.1 | |||
XM_006723728.3 | 1226 | Intron | XP_006723791.1 | |||
XM_006723730.3 | 1226 | Intron | XP_006723793.1 | |||
XM_006723732.2 | 1226 | Intron | XP_006723795.1 | |||
XM_006723733.1 | 1226 | Intron | XP_006723796.1 | |||
XM_011528648.2 | 1226 | Intron | XP_011526950.1 | |||
XM_011528652.2 | 1226 | UTR 5 | XP_011526954.1 | |||
XM_017027704.1 | 1226 | UTR 5 | XP_016883193.1 | |||
XM_017027705.1 | 1226 | Intron | XP_016883194.1 | |||
XM_017027706.1 | 1226 | Intron | XP_016883195.1 |