Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTAGTGTGTCTGTCAGGGAATGT[G/A]TCCCTCTTCCATTCTCTGTGTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
THEMIS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
THEMIS2 - thymocyte selection associated family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039477.2 | Intron | NP_001034566.1 | ||||
NM_001105556.2 | Intron | NP_001099026.1 | ||||
NM_001286113.1 | Intron | NP_001273042.1 | ||||
NM_001286115.1 | Intron | NP_001273044.1 | ||||
NM_004848.3 | Intron | NP_004839.2 | ||||
XM_005246041.3 | Intron | XP_005246098.1 | ||||
XM_006711050.1 | Intron | XP_006711113.1 | ||||
XM_011542445.2 | Intron | XP_011540747.1 |