Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAATTTCTACCAACCCAAATCCC[A/G]TTGGCATGGAACTCTGCACTGCAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF345 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF345 - zinc finger protein 345 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242472.1 | Intron | NP_001229401.1 | ||||
NM_001242474.1 | Intron | NP_001229403.1 | ||||
NM_001242475.1 | Intron | NP_001229404.1 | ||||
NM_001242476.1 | Intron | NP_001229405.1 | ||||
NM_003419.4 | Intron | NP_003410.1 | ||||
XM_011526704.2 | Intron | XP_011525006.1 | ||||
XM_017026568.1 | Intron | XP_016882057.1 | ||||
XM_017026569.1 | Intron | XP_016882058.1 | ||||
XM_017026570.1 | Intron | XP_016882059.1 | ||||
XM_017026571.1 | Intron | XP_016882060.1 | ||||
XM_017026572.1 | Intron | XP_016882061.1 | ||||
XM_017026573.1 | Intron | XP_016882062.1 |
Set Membership: |
HapMap |