Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTGTTAGGGTTAACACCTGGTC[C/T]GTGGAGGCATTCAGAAAGAATCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606443 MIM: 609471 MIM: 186745 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CREB3 - cAMP responsive element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006368.4 | Intron | NP_006359.3 |
GBA2 - glucosylceramidase beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6853 - microRNA 6853 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLN1 - talin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |