Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATACCGTTTCCTTTTGATCACAA[C/G]TGAAAAAGCATGTAATCGTTTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615211 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UHRF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UHRF2 - ubiquitin like with PHD and ring finger domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152896.2 | Intron | NP_690856.1 | ||||
XM_011517704.2 | Intron | XP_011516006.1 | ||||
XM_011517705.2 | Intron | XP_011516007.1 | ||||
XM_017014253.1 | Intron | XP_016869742.1 | ||||
XM_017014254.1 | Intron | XP_016869743.1 |
Set Membership: |
HapMap Validated |