Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600664 MIM: 616120 MIM: 611330 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHUK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHUK - conserved helix-loop-helix ubiquitous kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CWF19L1 - CWF19-like 1, cell cycle control (S. pombe) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303404.1 | Intron | NP_001290333.1 | ||||
NM_001303405.1 | Intron | NP_001290334.1 | ||||
NM_001303406.1 | Intron | NP_001290335.1 | ||||
NM_001303407.1 | Intron | NP_001290336.1 | ||||
NM_018294.5 | Intron | NP_060764.3 |
SNORA12 - small nucleolar RNA, H/ACA box 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |