Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTTTAATATTCTGGAGCTGGCT[A/G]TCTGGCACTCCCAATGTGCTGACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147582 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IREB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IREB2 - iron responsive element binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320941.1 | Intron | NP_001307870.1 | ||||
NM_001320942.1 | Intron | NP_001307871.1 | ||||
NM_001320943.1 | Intron | NP_001307872.1 | ||||
NM_004136.3 | Intron | NP_004127.1 |
Set Membership: |
HapMap |