Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTCTGCCTAGTCCTCATGTCCC[G/T]CCCCTCCCATCCTTGCCCGCTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604833 MIM: 606999 MIM: 600939 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL27 - C-C motif chemokine ligand 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GALT - galactose-1-phosphate uridylyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL11RA - interleukin 11 receptor subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142784.2 | Intron | NP_001136256.1 |
LOC730098 - uncharacterized LOC730098 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |