Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603944 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
STX6 - syntaxin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286210.1 | Intron | NP_001273139.1 | ||||
NM_005819.5 | Intron | NP_005810.1 | ||||
XM_011509058.1 | Intron | XP_011507360.1 | ||||
XM_011509059.1 | Intron | XP_011507361.1 | ||||
XM_017000006.1 | Intron | XP_016855495.1 | ||||
XM_017000007.1 | Intron | XP_016855496.1 |
Set Membership: |
HapMap Validated |