Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606525 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SRGAP3 - SLIT-ROBO Rho GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033117.2 | 8033 | UTR 3 | NP_001028289.1 | |||
NM_014850.3 | 8033 | UTR 3 | NP_055665.1 | |||
XM_011534295.1 | 8033 | UTR 3 | XP_011532597.1 | |||
XM_011534296.1 | 8033 | UTR 3 | XP_011532598.1 | |||
XM_011534301.2 | 8033 | UTR 3 | XP_011532603.1 | |||
XM_011534303.1 | 8033 | UTR 3 | XP_011532605.1 | |||
XM_017007574.1 | 8033 | UTR 3 | XP_016863063.1 | |||
XM_017007575.1 | 8033 | UTR 3 | XP_016863064.1 | |||
XM_017007576.1 | 8033 | UTR 3 | XP_016863065.1 | |||
XM_017007577.1 | 8033 | UTR 3 | XP_016863066.1 | |||
XM_017007578.1 | 8033 | UTR 3 | XP_016863067.1 | |||
XM_017007579.1 | 8033 | UTR 3 | XP_016863068.1 | |||
XM_017007580.1 | 8033 | Intron | XP_016863069.1 |
Set Membership: |
HapMap Validated |