Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSRC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RSRC2 - arginine and serine rich coiled-coil 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023012.5 | Intron | NP_075388.2 | ||||
XM_005253601.1 | Intron | XP_005253658.1 | ||||
XM_005253602.2 | Intron | XP_005253659.1 | ||||
XM_005253604.1 | Intron | XP_005253661.1 | ||||
XM_011538688.1 | Intron | XP_011536990.1 | ||||
XM_011538689.1 | Intron | XP_011536991.1 | ||||
XM_017019830.1 | Intron | XP_016875319.1 | ||||
XM_017019831.1 | Intron | XP_016875320.1 | ||||
XM_017019832.1 | Intron | XP_016875321.1 | ||||
XM_017019833.1 | Intron | XP_016875322.1 |