Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182115 MIM: 612543 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYTH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYTH1 - cytohesin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP36 - ubiquitin specific peptidase 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321291.1 | Intron | NP_001308220.1 | ||||
XM_005257542.2 | Intron | XP_005257599.1 | ||||
XM_005257543.2 | Intron | XP_005257600.1 | ||||
XM_011525065.1 | Intron | XP_011523367.1 | ||||
XM_011525066.2 | Intron | XP_011523368.1 | ||||
XM_011525067.2 | Intron | XP_011523369.1 | ||||
XM_011525068.1 | Intron | XP_011523370.1 | ||||
XM_011525069.2 | Intron | XP_011523371.1 | ||||
XM_011525070.1 | Intron | XP_011523372.1 | ||||
XM_011525071.2 | Intron | XP_011523373.1 | ||||
XM_017024900.1 | Intron | XP_016880389.1 | ||||
XM_017024901.1 | Intron | XP_016880390.1 |
Set Membership: |
HapMap |