Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTTTTTAAAATTGCTACACGA[A/C]AAAGCAGTTCCTTCTTTATTCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603061 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENSA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENSA - endosulfine alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004436.2 | Intron | NP_004427.1 | ||||
NM_207042.1 | Intron | NP_996925.1 | ||||
NM_207043.1 | Intron | NP_996926.1 | ||||
NM_207044.1 | Intron | NP_996927.1 | ||||
NM_207045.1 | Intron | NP_996928.1 | ||||
NM_207046.1 | Intron | NP_996929.1 | ||||
NM_207047.1 | Intron | NP_996930.1 | ||||
NM_207168.1 | Intron | NP_997051.1 |