Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGACTTACAATGGTTGGATTTGG[G/T]TGTGTGAAAGTAAATGTGTATTATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605862 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM5 - transmembrane protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278237.1 | Intron | NP_001265166.1 | ||||
NM_014254.2 | Intron | NP_055069.1 | ||||
XM_005268562.3 | Intron | XP_005268619.1 | ||||
XM_005268563.3 | Intron | XP_005268620.1 | ||||
XM_017018686.1 | Intron | XP_016874175.1 | ||||
XM_017018687.1 | Intron | XP_016874176.1 |
Set Membership: |
HapMap |