Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600365 MIM: 607251 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ABR - active BCR-related | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001092.4 | 3307 | UTR 3 | NP_001083.2 | |||
NM_001159746.2 | 3307 | UTR 3 | NP_001153218.1 | |||
NM_001256847.2 | 3307 | UTR 3 | NP_001243776.1 | |||
NM_001282149.1 | 3307 | UTR 3 | NP_001269078.1 | |||
NM_001322840.1 | 3307 | UTR 3 | NP_001309769.1 | |||
NM_001322841.1 | 3307 | UTR 3 | NP_001309770.1 | |||
NM_001322842.1 | 3307 | Intron | NP_001309771.1 | |||
NM_021962.4 | 3307 | UTR 3 | NP_068781.2 | |||
XM_011523810.2 | 3307 | UTR 3 | XP_011522112.1 | |||
XM_011523811.2 | 3307 | Intron | XP_011522113.2 | |||
XM_011523812.2 | 3307 | UTR 3 | XP_011522114.1 | |||
XM_011523813.2 | 3307 | UTR 3 | XP_011522115.1 | |||
XM_011523814.2 | 3307 | UTR 3 | XP_011522116.1 | |||
XM_011523815.2 | 3307 | UTR 3 | XP_011522117.1 | |||
XM_017024539.1 | 3307 | UTR 3 | XP_016880028.1 | |||
XM_017024540.1 | 3307 | UTR 3 | XP_016880029.1 | |||
XM_017024541.1 | 3307 | Intron | XP_016880030.1 | |||
XM_017024542.1 | 3307 | UTR 3 | XP_016880031.1 | |||
XM_017024543.1 | 3307 | UTR 3 | XP_016880032.1 |
TIMM22 - translocase of inner mitochondrial membrane 22 homolog (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |