Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGCTGAGCAGGAGGAATAGAG[A/T]GGAAAATGAGAAAAAGGAAACAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 122561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRHR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRHR1 - corticotropin releasing hormone receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145146.1 | Intron | NP_001138618.1 | ||||
NM_001145147.1 | Intron | NP_001138619.1 | ||||
NM_001145148.1 | Intron | NP_001138620.1 | ||||
NM_001303018.1 | Intron | NP_001289947.1 | ||||
NM_001303020.1 | Intron | NP_001289949.1 | ||||
NM_004382.4 | Intron | NP_004373.2 |
MGC57346-CRHR1 - MGC57346-CRHR1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256299.2 | Intron | NP_001243228.1 | ||||
NM_001303016.1 | Intron | NP_001289945.1 |