Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTAAAATGAAAAATATGTTGCT[A/G]GCTTTTTTTGAAGTTGGTTTTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607686 MIM: 609732 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FIP1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FIP1L1 - factor interacting with PAPOLA and CPSF1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LNX1 - ligand of numb-protein X 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126328.2 | Intron | NP_001119800.1 | ||||
NM_032622.2 | Intron | NP_116011.2 | ||||
XM_005265785.4 | Intron | XP_005265842.1 | ||||
XM_017008776.1 | Intron | XP_016864265.1 |
Set Membership: |
HapMap |