Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACAAATGAATTCTGAAGGGAC[A/G]CATAAGACATTGGTATTGGTTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929762 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101929762 - uncharacterized LOC101929762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP53 - ubiquitin specific peptidase 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019050.2 | Intron | NP_061923.2 | ||||
XM_005263073.3 | Intron | XP_005263130.1 | ||||
XM_005263075.2 | Intron | XP_005263132.1 | ||||
XM_005263076.2 | Intron | XP_005263133.1 | ||||
XM_005263077.3 | Intron | XP_005263134.1 | ||||
XM_006714240.1 | Intron | XP_006714303.1 | ||||
XM_011532037.2 | Intron | XP_011530339.1 | ||||
XM_011532038.2 | Intron | XP_011530340.1 | ||||
XM_017008312.1 | Intron | XP_016863801.1 | ||||
XM_017008313.1 | Intron | XP_016863802.1 | ||||
XM_017008314.1 | Intron | XP_016863803.1 | ||||
XM_017008315.1 | Intron | XP_016863804.1 | ||||
XM_017008316.1 | Intron | XP_016863805.1 | ||||
XM_017008317.1 | Intron | XP_016863806.1 | ||||
XM_017008318.1 | Intron | XP_016863807.1 |
Set Membership: |
Validated |