Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605494 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB3BP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITGB3BP - integrin subunit beta 3 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206739.1 | 1012 | Intron | NP_001193668.1 | |||
NM_014288.4 | 1012 | Intron | NP_055103.3 | |||
XM_006710514.3 | 1012 | Intron | XP_006710577.3 | |||
XM_006710515.3 | 1012 | UTR 3 | XP_006710578.3 | |||
XM_017000853.1 | 1012 | Intron | XP_016856342.1 | |||
XM_017000854.1 | 1012 | Intron | XP_016856343.1 | |||
XM_017000855.1 | 1012 | Intron | XP_016856344.1 | |||
XM_017000856.1 | 1012 | Intron | XP_016856345.1 | |||
XM_017000857.1 | 1012 | Intron | XP_016856346.1 |