Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611122 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD28 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD28 - ankyrin repeat domain 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195098.1 | Intron | NP_001182027.1 | ||||
NM_001195099.1 | Intron | NP_001182028.1 | ||||
NM_015199.3 | Intron | NP_056014.2 | ||||
XM_005264996.3 | Intron | XP_005265053.1 | ||||
XM_011533540.2 | Intron | XP_011531842.1 | ||||
XM_011533541.2 | Intron | XP_011531843.1 | ||||
XM_011533542.2 | Intron | XP_011531844.1 | ||||
XM_011533543.2 | Intron | XP_011531845.1 | ||||
XM_011533545.2 | Intron | XP_011531847.1 | ||||
XM_011533546.2 | Intron | XP_011531848.1 | ||||
XM_011533547.2 | Intron | XP_011531849.1 | ||||
XM_017006026.1 | Intron | XP_016861515.1 | ||||
XM_017006027.1 | Intron | XP_016861516.1 | ||||
XM_017006028.1 | Intron | XP_016861517.1 | ||||
XM_017006029.1 | Intron | XP_016861518.1 |
Set Membership: |
HapMap |