Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613572 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPRC6A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPRC6A - G protein-coupled receptor class C group 6 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286354.1 | 7115 | Intron | NP_001273283.1 | |||
NM_001286355.1 | 7115 | Intron | NP_001273284.1 | |||
NM_148963.3 | 7115 | Intron | NP_683766.2 | |||
XM_017010475.1 | 7115 | UTR 3 | XP_016865964.1 | |||
XM_017010476.1 | 7115 | Intron | XP_016865965.1 |
Set Membership: |
HapMap |