Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614169 | ||||||||||||||||||||
Literature Links: |
CCDC12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC12 - coiled-coil domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277074.1 | Intron | NP_001264003.1 | ||||
NM_144716.5 | Intron | NP_653317.2 | ||||
XM_011533390.2 | Intron | XP_011531692.1 | ||||
XM_011533391.2 | Intron | XP_011531693.1 | ||||
XM_011533392.2 | Intron | XP_011531694.1 | ||||
XM_011533393.2 | Intron | XP_011531695.1 | ||||
XM_011533394.2 | Intron | XP_011531696.1 | ||||
XM_011533396.2 | Intron | XP_011531698.1 |
NBEAL2 - neurobeachin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015175.2 | Intron | NP_055990.1 | ||||
XM_006713072.3 | Intron | XP_006713135.1 | ||||
XM_011533533.2 | Intron | XP_011531835.1 | ||||
XM_017006010.1 | Intron | XP_016861499.1 | ||||
XM_017006011.1 | Intron | XP_016861500.1 | ||||
XM_017006012.1 | Intron | XP_016861501.1 | ||||
XM_017006013.1 | Intron | XP_016861502.1 | ||||
XM_017006014.1 | Intron | XP_016861503.1 | ||||
XM_017006015.1 | Intron | XP_016861504.1 | ||||
XM_017006016.1 | Intron | XP_016861505.1 | ||||
XM_017006017.1 | Intron | XP_016861506.1 |