Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAAGCACCATGGCAGACTTATGC[C/T]CTGGCTTTAGAAGCCTGTATGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600579 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRO PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PTPRO - protein tyrosine phosphatase, receptor type O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002848.3 | Intron | NP_002839.1 | ||||
NM_030667.2 | Intron | NP_109592.1 | ||||
NM_030668.2 | Intron | NP_109593.1 | ||||
NM_030669.2 | Intron | NP_109594.1 | ||||
NM_030670.2 | Intron | NP_109595.1 | ||||
NM_030671.2 | Intron | NP_109596.1 | ||||
XM_017019725.1 | Intron | XP_016875214.1 |
Set Membership: |
HapMap Validated |