Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTTCACCTGCCCAGCCCCAGCC[T/C]GCTGTCACCGGGGTAGGTTACGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612081 MIM: 605592 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL34 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL34 - interleukin 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172771.1 | Intron | NP_001166242.1 | ||||
NM_001172772.1 | Intron | NP_001166243.1 | ||||
NM_152456.2 | Intron | NP_689669.2 | ||||
XM_011522902.2 | Intron | XP_011521204.1 | ||||
XM_011522903.2 | Intron | XP_011521205.1 |
SF3B3 - splicing factor 3b subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |