Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGGCAAACATGATTACTACTAC[A/G]CTATGAGAACAGACTCTCTCATTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604875 MIM: 608659 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO9A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MYO9A - myosin IXA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SENP8 - SUMO/sentrin peptidase family member, NEDD8 specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166340.1 | Intron | NP_001159812.1 | ||||
NM_001172109.1 | Intron | NP_001165580.1 | ||||
NM_001172110.1 | Intron | NP_001165581.1 | ||||
NM_001172111.1 | Intron | NP_001165582.1 | ||||
NM_145204.3 | Intron | NP_660205.3 | ||||
XM_005254157.3 | Intron | XP_005254214.1 | ||||
XM_011521215.2 | Intron | XP_011519517.1 | ||||
XM_011521217.2 | Intron | XP_011519519.1 |
Set Membership: |
HapMap Validated |