Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAGGACTGCAGGAAGCAAAGTA[C/T]GGAGGGAGGAAAGGCATTACTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606686 MIM: 601750 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EIF2B1 - eukaryotic translation initiation factor 2B subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GTF2H3 - general transcription factor IIH subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271866.1 | Intron | NP_001258795.1 | ||||
NM_001271867.1 | Intron | NP_001258796.1 | ||||
NM_001271868.1 | Intron | NP_001258797.1 | ||||
NM_001516.4 | Intron | NP_001507.2 | ||||
XM_017019228.1 | Intron | XP_016874717.1 |
Set Membership: |
HapMap |