Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607809 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACAT1 - acetyl-CoA acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000019.3 | 74 | Missense Mutation | CCG,GCG | P,A 5 | NP_000010.1 | |
XM_006718835.3 | 74 | Intron | XP_006718898.1 | |||
XM_017017681.1 | 74 | UTR 5 | XP_016873170.1 | |||
XM_017017682.1 | 74 | UTR 5 | XP_016873171.1 | |||
XM_017017683.1 | 74 | UTR 5 | XP_016873172.1 |
Set Membership: |
HapMap |