Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613868 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC14A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC14A1 - solute carrier family 14 member 1 (Kidd blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128588.3 | Intron | NP_001122060.3 | ||||
NM_001146036.2 | Intron | NP_001139508.2 | ||||
NM_001146037.1 | Intron | NP_001139509.1 | ||||
NM_001308278.1 | Intron | NP_001295207.1 | ||||
NM_001308279.1 | Intron | NP_001295208.1 | ||||
NM_015865.6 | Intron | NP_056949.4 | ||||
XM_005258329.1 | Intron | XP_005258386.1 | ||||
XM_005258333.1 | Intron | XP_005258390.1 | ||||
XM_006722526.3 | Intron | XP_006722589.1 | ||||
XM_011526142.1 | Intron | XP_011524444.1 | ||||
XM_011526144.1 | Intron | XP_011524446.1 |
Set Membership: |
HapMap Validated |