Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
64 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600222 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TIE1 - tyrosine kinase with immunoglobulin like and EGF like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253357.1 | Intron | NP_001240286.1 | ||||
NM_005424.4 | Intron | NP_005415.1 | ||||
XM_005271163.1 | Intron | XP_005271220.1 | ||||
XM_006710869.1 | Intron | XP_006710932.1 | ||||
XM_017002207.1 | Intron | XP_016857696.1 |
Set Membership: |
HapMap |