Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGCTGTGGCCAAAAGCTTTTC[C/T]GCTGAACTTCCTGCTGTGCCCAGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610389 MIM: 612942 MIM: 605440 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAMTOR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LAMTOR2 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145264.1 | Intron | NP_001138736.1 | ||||
NM_014017.3 | Intron | NP_054736.1 |
RAB25 - RAB25, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBQLN4 - ubiquilin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |