Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601890 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KLC4 - kinesin light chain 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTK7 - protein tyrosine kinase 7 (inactive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270398.1 | Intron | NP_001257327.1 | ||||
NM_002821.4 | Intron | NP_002812.2 | ||||
NM_152880.3 | Intron | NP_690619.1 | ||||
NM_152881.3 | Intron | NP_690620.1 | ||||
NM_152882.3 | Intron | NP_690621.1 | ||||
XM_011514765.2 | Intron | XP_011513067.1 | ||||
XM_011514766.2 | Intron | XP_011513068.1 |
Set Membership: |
HapMap Validated |