Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611298 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf194 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C1orf194 - chromosome 1 open reading frame 194 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA1324 - KIAA1324 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267048.1 | Intron | NP_001253977.1 | ||||
NM_001284352.1 | Intron | NP_001271281.1 | ||||
NM_001284353.1 | Intron | NP_001271282.1 | ||||
NM_020775.4 | Intron | NP_065826.2 | ||||
XM_011541825.1 | Intron | XP_011540127.1 | ||||
XM_011541826.2 | Intron | XP_011540128.1 | ||||
XM_011541827.1 | Intron | XP_011540129.1 | ||||
XM_017001883.1 | Intron | XP_016857372.1 |