Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTTCAGACTCACCTGGGGTGGG[A/G]GTACATTCAACTACATGGACGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300890 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASB9 - ankyrin repeat and SOCS box containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031739.2 | Intron | NP_001026909.1 | ||||
NM_001168530.1 | Intron | NP_001162002.1 | ||||
NM_001168531.1 | Intron | NP_001162003.1 | ||||
NM_024087.2 | Intron | NP_076992.1 | ||||
XM_005274446.1 | Intron | XP_005274503.1 | ||||
XM_011545458.1 | Intron | XP_011543760.1 | ||||
XM_011545460.1 | Intron | XP_011543762.1 | ||||
XM_017029283.1 | Intron | XP_016884772.1 | ||||
XM_017029284.1 | Intron | XP_016884773.1 | ||||
XM_017029285.1 | Intron | XP_016884774.1 | ||||
XM_017029286.1 | Intron | XP_016884775.1 |