Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTAAGGAGCATAAGATGTCCCTA[T/C]GTAAAATGCTAGTGTAGCATACAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123812 MIM: 603135 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CUL2 - cullin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |