Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCTCATCCAGGGGCTGAGAGA[C/T]GGCGGGACGCTGGGGCAGGGCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf129 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf129 - chromosome 9 open reading frame 129 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098808.1 | 936 | Missense Mutation | CAT,CGT | H,R 191 | NP_001092278.1 | |
XM_006717117.3 | 936 | Missense Mutation | CAT,CGT | H,R 191 | XP_006717180.1 |