Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGCAAAAGAACCAGGCCCTTT[A/C]TGCCTGGAGCTGTCACTCCTGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171840 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927824 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101927824 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFKP - phosphofructokinase, platelet | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242339.1 | Intron | NP_001229268.1 | ||||
NM_001323067.1 | Intron | NP_001309996.1 | ||||
NM_001323068.1 | Intron | NP_001309997.1 | ||||
NM_001323069.1 | Intron | NP_001309998.1 | ||||
NM_001323070.1 | Intron | NP_001309999.1 | ||||
NM_001323071.1 | Intron | NP_001310000.1 | ||||
NM_001323072.1 | Intron | NP_001310001.1 | ||||
NM_001323073.1 | Intron | NP_001310002.1 | ||||
NM_001323074.1 | Intron | NP_001310003.1 | ||||
NM_002627.4 | Intron | NP_002618.1 | ||||
XM_005252465.4 | Intron | XP_005252522.1 | ||||
XM_005252466.4 | Intron | XP_005252523.1 | ||||
XM_005252467.2 | Intron | XP_005252524.1 | ||||
XM_006717449.1 | Intron | XP_006717512.1 |
Set Membership: |
Validated |