Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCTCCCGACCCCATTACCCTC[C/T]ATGTGATGTATTTTTGGTTGATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616731 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984562 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107984562 - serine/threonine-protein kinase Nek5-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK5 - NIMA related kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199289.2 | Intron | NP_954983.1 | ||||
XM_006719808.3 | Intron | XP_006719871.2 | ||||
XM_011535066.2 | Intron | XP_011533368.1 | ||||
XM_011535068.2 | Intron | XP_011533370.1 | ||||
XM_017020555.1 | Intron | XP_016876044.1 | ||||
XM_017020556.1 | Intron | XP_016876045.1 | ||||
XM_017020557.1 | Intron | XP_016876046.1 |
Set Membership: |
HapMap |