Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFBR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGFBR2 - transforming growth factor beta receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024847.2 | 219 | Intron | NP_001020018.1 | |||
NM_003242.5 | 219 | Intron | NP_003233.4 | |||
XM_011534043.2 | 219 | Missense Mutation | AGT,GGT | S,G 6 | XP_011532345.1 | |
XM_011534045.2 | 219 | Intron | XP_011532347.1 | |||
XM_017007106.1 | 219 | Intron | XP_016862595.1 |