Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCGCATGCGCATCACCTGGGCG[G/T]CACCCGCGGTACTGCGCCTGCGCGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176264 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNC3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCNC3 - potassium voltage-gated channel subfamily C member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004977.2 | Intron | NP_004968.2 | ||||
XM_006723203.2 | Intron | XP_006723266.1 | ||||
XM_011526925.2 | Intron | XP_011525227.1 | ||||
XM_011526926.1 | Intron | XP_011525228.1 | ||||
XM_011526928.2 | Intron | XP_011525230.1 |
NAPSB - napsin B aspartic peptidase, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |